Assay Details
| Gene Symbol: NAC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113825.4 | NP_189546.1 | 2 - 3 | 666 | 113 | |
| GenBank mRNA | AY062638.1 | - | 2 - 3 | 619 | 113 | |
| BT008716.1 | - | 2 - 3 | 492 | 113 | ||
More Information
Set Membership: |
|

NAC3
Genomic Map