Assay Details
| Gene Symbol: CM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113844.4 | NP_566846.1 | 2 - 3 | 634 | 93 | |
| GenBank mRNA | BT024732.1 | - | 2 - 3 | 434 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CM1| Gene Symbol: CM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113844.4 | NP_566846.1 | 2 - 3 | 634 | 93 | |
| GenBank mRNA | BT024732.1 | - | 2 - 3 | 434 | 93 | |
Set Membership: |
|