Assay Details
| Gene Symbol: MYB121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113920.2 | NP_189640.1 | 1 - 2 | 379 | 74 | |
| GenBank mRNA | AF371980.2 | - | 1 - 2 | 379 | 74 | |
| AY519593.1 | - | 1 - 2 | 175 | 74 | ||
More Information
Set Membership: |
|

MYB121
Genomic Map