Assay Details
| Gene Symbol: MYB121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113920.2 | NP_189640.1 | 2 - 3 | 514 | 118 | |
| GenBank mRNA | AF371980.2 | - | 2 - 3 | 514 | 118 | |
| AY519593.1 | - | 2 - 3 | 310 | 118 | ||
More Information
Set Membership: |
|

MYB121
Genomic Map