Assay Details
| Gene Symbol: AGL79 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113925.3 | NP_189645.2 | 1 - 2 | 432 | 106 | |
| GenBank mRNA | AY141238.1 | - | 1 - 2 | 183 | 106 | |
More Information
Set Membership: |
|

AGL79
Genomic Map