Assay Details
| Gene Symbol: AGL79 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113925.3 | NP_189645.2 | 5 - 6 | 725 | 78 | |
| GenBank mRNA | AY141238.1 | - | 5 - 6 | 476 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AGL79| Gene Symbol: AGL79 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113925.3 | NP_189645.2 | 5 - 6 | 725 | 78 | |
| GenBank mRNA | AY141238.1 | - | 5 - 6 | 476 | 78 | |
Set Membership: |
|