Assay Details
| Gene Symbol: AT3G45450 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114414.1 | NP_190131.1 | 4 - 5 | 455 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT3G45450| Gene Symbol: AT3G45450 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114414.1 | NP_190131.1 | 4 - 5 | 455 | 62 | |
Set Membership: |
|