Assay Details
| Gene Symbol: NAXT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339214.1 | NP_001327486.1 | 2 - 3 | 691 | 99 | |
| NM_114434.2 | NP_190151.1 | 2 - 3 | 691 | 99 | ||
More Information
Set Membership: |
|

NAXT1
Genomic Map