Assay Details
| Gene Symbol: HRS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114479.5 | NP_190196.1 | 1 - 2 | 356 | 89 | |
| GenBank mRNA | AF020715.1 | - | 1 - 2 | 292 | 89 | |
| AY056233.1 | - | 1 - 2 | 297 | 89 | ||
| AY117252.1 | - | 1 - 2 | 287 | 89 | ||
More Information
Set Membership: |
|

HRS1
Genomic Map