Assay Details
| Gene Symbol: HRS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114479.5 | NP_190196.1 | 5 - 6 | 762 | 78 | |
| GenBank mRNA | AF020715.1 | - | 5 - 6 | 698 | 78 | |
| AY056233.1 | - | 5 - 6 | 703 | 78 | ||
| AY117252.1 | - | 5 - 6 | 693 | 78 | ||
More Information
Set Membership: |
|

HRS1
Genomic Map