Assay Details
| Gene Symbol: HRS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114479.5 | NP_190196.1 | 9 - 10 | 1232 | 63 | |
| GenBank mRNA | AF020715.1 | - | 9 - 10 | 1168 | 63 | |
| AY056233.1 | - | 9 - 10 | 1173 | 63 | ||
| AY117252.1 | - | 9 - 10 | 1163 | 63 | ||
More Information
Set Membership: |
|

HRS1
Genomic Map