Assay Details
| Gene Symbol: MBD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114525.3 | NP_190242.1 | 2 - 3 | 460 | 60 | |
| GenBank mRNA | AY063849.1 | - | 2 - 3 | 350 | 60 | |
| AY142653.1 | - | 2 - 3 | 295 | 60 | ||
More Information
Set Membership: |
|

MBD5
Genomic Map