Assay Details
| Gene Symbol: TRFL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114526.3 | NP_190243.2 | 2 - 3 | 461 | 92 | |
| GenBank mRNA | AJ630475.1 | - | 1 - 2 | 85 | 92 | |
| AY519541.1 | - | 1 - 2 | 85 | 92 | ||
| AY568647.1 | - | 1 - 2 | 85 | 92 | ||
More Information
Set Membership: |
|

TRFL1
Genomic Map