Assay Details
| Gene Symbol: TOC75-III | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114541.3 | NP_190258.1 | 1 - 2 | 1305 | 96 | |
| GenBank mRNA | AY127014.1 | - | 1 - 2 | 1237 | 96 | |
| BT006358.1 | - | 1 - 2 | 1190 | 96 | ||
More Information
Set Membership: |
|

TOC75-III
Genomic Map