Assay Details
| Gene Symbol: TOC75-III | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114541.3 | NP_190258.1 | 2 - 3 | 1531 | 100 | |
| GenBank mRNA | AY127014.1 | - | 2 - 3 | 1463 | 100 | |
| BT006358.1 | - | 2 - 3 | 1416 | 100 | ||
More Information
Set Membership: |
|

TOC75-III
Genomic Map