Assay Details
| Gene Symbol: LHCA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114615.4 | NP_190331.3 | 1 - 2 | 471 | 88 | |
| GenBank mRNA | AY086470.1 | - | 1 - 2 | 472 | 88 | |
| AY093080.1 | - | 1 - 2 | 311 | 88 | ||
| BT000093.1 | - | 1 - 2 | 211 | 88 | ||
| M63931.1 | - | 1 - 2 | 281 | 88 | ||
| Gene Symbol: VRN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK230388.1 | - | 1730 | 88 | ||
More Information
Set Membership: |
|

LHCA4
Genomic Map