Assay Details
| Gene Symbol: LHCA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114615.4 | NP_190331.3 | 3 - 4 | 888 | 67 | |
| GenBank mRNA | AY086470.1 | - | 3 - 4 | 889 | 67 | |
| AY093080.1 | - | 3 - 4 | 728 | 67 | ||
| BT000093.1 | - | 3 - 4 | 628 | 67 | ||
| M63931.1 | - | 3 - 4 | 698 | 67 | ||
| Gene Symbol: VRN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK230388.1 | - | 2147 | 67 | ||
More Information
Set Membership: |
|

LHCA4
Genomic Map