Assay Details
| Gene Symbol: MYB94 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339319.1 | NP_001325642.1 | 1 - 2 | 798 | 91 | |
| NM_114628.4 | NP_190344.1 | 2 - 3 | 539 | 91 | ||
| GenBank mRNA | AY519595.1 | - | 2 - 3 | 267 | 91 | |
| BT002802.1 | - | 2 - 3 | 336 | 91 | ||
| BT004361.1 | - | 2 - 3 | 267 | 91 | ||
More Information
Set Membership: |
|

MYB94
Genomic Map