Assay Details
| Gene Symbol: EB1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114637.4 | NP_190353.3 | 5 - 6 | 771 | 64 | |
| GenBank mRNA | AK227867.1 | - | 5 - 6 | 736 | 64 | |
| BT009707.1 | - | 5 - 6 | 514 | 64 | ||
More Information
Set Membership: |
|

EB1a
Genomic Map