Assay Details
| Gene Symbol: ALDH2B4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114669.4 | NP_190383.1 | 10 - 11 | 1670 | 61 | |
| GenBank mRNA | AB030820.1 | - | 10 - 11 | 1426 | 61 | |
| AF327426.1 | - | 10 - 11 | 1420 | 61 | ||
| AF349447.1 | - | 10 - 11 | 1426 | 61 | ||
| AF349522.1 | - | 10 - 11 | 1344 | 61 | ||
| AF372911.1 | - | 10 - 11 | 1420 | 61 | ||
| AY090443.1 | - | 10 - 11 | 1424 | 61 | ||
| BT000752.1 | - | 10 - 11 | 1369 | 61 | ||
| BT006371.1 | - | 10 - 11 | 1344 | 61 | ||
More Information
Set Membership: |
|

ALDH2B4
Genomic Map