Assay Details
| Gene Symbol: ALDH2B4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114669.4 | NP_190383.1 | 7 - 8 | 1330 | 112 | |
| GenBank mRNA | AB030820.1 | - | 7 - 8 | 1086 | 112 | |
| AF327426.1 | - | 7 - 8 | 1080 | 112 | ||
| AF349447.1 | - | 7 - 8 | 1086 | 112 | ||
| AF349522.1 | - | 7 - 8 | 1004 | 112 | ||
| AF372911.1 | - | 7 - 8 | 1080 | 112 | ||
| AY090443.1 | - | 7 - 8 | 1084 | 112 | ||
| BT000752.1 | - | 7 - 8 | 1029 | 112 | ||
| BT006371.1 | - | 7 - 8 | 1004 | 112 | ||
More Information
Set Membership: |
|

ALDH2B4
Genomic Map