Assay Details
| Gene Symbol: APC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339352.1 | NP_001326941.1 | 1 - 2 | 365 | 106 | |
| NM_114684.5 | NP_190398.1 | 2 - 3 | 515 | 106 | ||
| GenBank mRNA | AY057562.1 | - | 2 - 3 | 478 | 106 | |
| AY099779.1 | - | 2 - 3 | 478 | 106 | ||
| BT000289.1 | - | 2 - 3 | 398 | 106 | ||
More Information
Set Membership: |
|

APC8
Genomic Map