Assay Details
| Gene Symbol: BT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339366.1 | NP_001326127.1 | 1 - 2 | 428 | 85 | |
| NM_001339367.1 | NP_001326126.1 | 1 - 2 | 426 | 85 | ||
| NM_114697.4 | NP_566902.1 | 1 - 2 | 428 | 85 | ||
| GenBank mRNA | AK226596.1 | - | 1 - 2 | 216 | 85 | |
| AY040015.1 | - | 1 - 2 | 214 | 85 | ||
| AY079408.1 | - | 1 - 2 | 161 | 85 | ||
| AY316675.1 | - | 1 - 2 | 161 | 85 | ||
| BT000678.1 | - | 1 - 2 | 207 | 85 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001339367.1 transcripts.
Set Membership: |
|

BT2
Genomic Map