Assay Details
| Gene Symbol: BT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339366.1 | NP_001326127.1 | 2 - 3 | 570 | 94 | |
| NM_114697.4 | NP_566902.1 | 2 - 3 | 570 | 94 | ||
| GenBank mRNA | AK226596.1 | - | 2 - 3 | 358 | 94 | |
| AY040015.1 | - | 2 - 3 | 356 | 94 | ||
| AY079408.1 | - | 2 - 3 | 303 | 94 | ||
| AY316675.1 | - | 2 - 3 | 303 | 94 | ||
| BT000678.1 | - | 2 - 3 | 349 | 94 | ||
More Information
Set Membership: |
|

BT2
Genomic Map