Assay Details
| Gene Symbol: BT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339366.1 | NP_001326127.1 | 3 - 4 | 875 | 65 | |
| NM_001339367.1 | NP_001326126.1 | 2 - 3 | 1077 | 65 | ||
| NM_114697.4 | NP_566902.1 | 3 - 4 | 875 | 65 | ||
| GenBank mRNA | AK226596.1 | - | 3 - 4 | 663 | 65 | |
| AY040015.1 | - | 3 - 4 | 661 | 65 | ||
| AY079408.1 | - | 3 - 4 | 608 | 65 | ||
| AY316675.1 | - | 3 - 4 | 608 | 65 | ||
| BT000678.1 | - | 3 - 4 | 654 | 65 | ||
More Information
Set Membership: |
|

BT2
Genomic Map