Assay Details
| Gene Symbol: AT3G48540 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114712.3 | NP_190423.2 | 7 - 8 | 546 | 101 | |
| GenBank mRNA | DQ446745.1 | - | 7 - 8 | 522 | 101 | |
More Information
Set Membership: |
|

AT3G48540
Genomic Map