Assay Details
| Gene Symbol: CDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114734.4 | NP_566911.1 | 6 - 7 | 922 | 68 | |
| GenBank mRNA | AK226373.1 | - | 6 - 7 | 820 | 68 | |
| AY085153.1 | - | 6 - 7 | 822 | 68 | ||
| BT024706.1 | - | 4 - 5 | 653 | 68 | ||
| M59198.1 | - | 6 - 7 | 820 | 68 | ||
| S45387.1 | - | 6 - 7 | 820 | 68 | ||
| X57839.1 | - | 811 | 68 | |||
More Information
Set Membership: |
|

CDC2
Genomic Map