Assay Details
| Gene Symbol: AT3G48830 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339381.1 | NP_001319712.1 | 5 - 6 | 577 | 139 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT3G48830| Gene Symbol: AT3G48830 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339381.1 | NP_001319712.1 | 5 - 6 | 577 | 139 | |
Set Membership: |
|