Assay Details
| Gene Symbol: ABIL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035755.2 | NP_001030832.1 | 2 - 3 | 408 | 92 | |
| NM_114788.4 | NP_190498.1 | 2 - 3 | 661 | 92 | ||
| GenBank mRNA | AY817013.1 | - | 1 - 2 | 73 | 92 | |
| BT026081.1 | - | 1 - 2 | 73 | 92 | ||
More Information
Set Membership: |
|

ABIL2
Genomic Map