Assay Details
| Gene Symbol: SR34a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084795.1 | NP_001078264.1 | 11 - 12 | 1182 | 69 | |
| NM_001203112.1 | NP_001190041.1 | 13 - 14 | 1323 | 69 | ||
| NM_001339417.1 | NP_001325875.1 | 11 - 12 | 2037 | 69 | ||
| NM_001339418.1 | NP_001325874.1 | 12 - 13 | 1867 | 69 | ||
| NM_001339419.1 | NP_001325876.1 | 12 - 13 | 1502 | 69 | ||
| NM_001339420.1 | NP_001325877.1 | 13 - 14 | 1295 | 69 | ||
| NM_114803.6 | NP_190512.3 | 12 - 13 | 1108 | 69 | ||
More Information
Set Membership: |
|

SR34a
Genomic Map