Assay Details
| Gene Symbol: SR34a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084795.1 | NP_001078264.1 | 4 - 5 | 422 | 69 | |
| NM_001203112.1 | NP_001190041.1 | 5 - 6 | 674 | 69 | ||
| NM_001339417.1 | NP_001325875.1 | 3 - 4 | 1388 | 69 | ||
| NM_001339418.1 | NP_001325874.1 | 4 - 5 | 1218 | 69 | ||
| NM_001339419.1 | NP_001325876.1 | 4 - 5 | 853 | 69 | ||
| NM_001339420.1 | NP_001325877.1 | 5 - 6 | 646 | 69 | ||
| NM_114803.6 | NP_190512.3 | 4 - 5 | 459 | 69 | ||
| GenBank mRNA | BT030067.1 | - | 3 - 4 | 229 | 69 | |
More Information
Set Membership: |
|

SR34a
Genomic Map