Assay Details
| Gene Symbol: UBP26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339429.1 | NP_001325635.1 | 5 - 6 | 1842 | 101 | |
| NM_001339430.1 | NP_001325634.1 | 6 - 7 | 1793 | 101 | ||
| NM_114820.4 | NP_566922.1 | 5 - 6 | 1953 | 101 | ||
More Information
Set Membership: |
|

UBP26
Genomic Map