Assay Details
| Gene Symbol: DIN11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114822.5 | NP_190531.2 | 11 - 12 | 996 | 145 | |
| GenBank mRNA | BT000898.1 | - | 11 - 12 | 920 | 145 | |
More Information
Set Membership: |
|

DIN11
Genomic Map