Assay Details
| Gene Symbol: TRB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339442.1 | NP_001326904.1 | 3 - 4 | 861 | 66 | |
| NM_114845.5 | NP_190554.1 | 4 - 5 | 716 | 66 | ||
| GenBank mRNA | AY058887.1 | - | 4 - 5 | 656 | 66 | |
| AY065069.1 | - | 4 - 5 | 660 | 66 | ||
| AY079043.1 | - | 3 - 4 | 444 | 66 | ||
| AY519542.1 | - | 3 - 4 | 444 | 66 | ||
| U83839.1 | - | 4 - 5 | 664 | 66 | ||
More Information
Set Membership: |
|

TRB3
Genomic Map