Assay Details
Gene Symbol: TRB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339442.1 | NP_001326904.1 | 4 - 5 | 949 | 120 | |
NM_114845.5 | NP_190554.1 | 5 - 6 | 804 | 120 | ||
GenBank mRNA | AY058887.1 | - | 5 - 6 | 744 | 120 | |
AY065069.1 | - | 5 - 6 | 748 | 120 | ||
AY079043.1 | - | 4 - 5 | 532 | 120 | ||
AY519542.1 | - | 4 - 5 | 532 | 120 | ||
U83839.1 | - | 5 - 6 | 752 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
