Assay Details
| Gene Symbol: HR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339471.1 | NP_001325500.1 | 1 - 2 | 573 | 94 | |
| NM_114908.5 | NP_566933.1 | 1 - 2 | 552 | 94 | ||
| GenBank mRNA | AF361849.1 | - | 1 - 2 | 418 | 94 | |
| BT000848.1 | - | 1 - 2 | 307 | 94 | ||
More Information
Set Membership: |
|

HR4
Genomic Map