Assay Details
| Gene Symbol: AT3G50520 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114912.5 | NP_190621.1 | 2 - 3 | 202 | 85 | |
| GenBank mRNA | AY035101.1 | - | 2 - 3 | 127 | 85 | |
| AY063062.1 | - | 2 - 3 | 98 | 85 | ||
More Information
Set Membership: |
|

AT3G50520
Genomic Map