Assay Details
| Gene Symbol: AT3G50520 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114912.5 | NP_190621.1 | 4 - 5 | 362 | 71 | |
| GenBank mRNA | AY035101.1 | - | 4 - 5 | 287 | 71 | |
| AY063062.1 | - | 4 - 5 | 258 | 71 | ||
More Information
Set Membership: |
|

AT3G50520
Genomic Map