Assay Details
| Gene Symbol: DWF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339479.1 | NP_001327730.1 | 4 - 5 | 1254 | 67 | |
| NM_114926.4 | NP_190635.1 | 4 - 5 | 1254 | 67 | ||
| GenBank mRNA | AF412114.1 | - | 4 - 5 | 1045 | 67 | |
| AY090266.1 | - | 4 - 5 | 1017 | 67 | ||
More Information
Set Membership: |
|

DWF4
Genomic Map