Assay Details
| Gene Symbol: DWF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114926.4 | NP_190635.1 | 7 - 8 | 1535 | 135 | |
| GenBank mRNA | AF412114.1 | - | 7 - 8 | 1326 | 135 | |
| AY090266.1 | - | 7 - 8 | 1298 | 135 | ||
More Information
Set Membership: |
|

DWF4
Genomic Map