Assay Details
| Gene Symbol: AT3G50810 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339482.1 | NP_001326370.1 | 2 - 3 | 377 | 120 | |
| NM_001339483.1 | NP_001326371.1 | 4 - 5 | 348 | 120 | ||
| NM_114941.5 | NP_190650.4 | 3 - 4 | 402 | 120 | ||
More Information
Set Membership: |
|

AT3G50810
Genomic Map