Assay Details
| Gene Symbol: AT3G50910 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114951.3 | NP_566941.1 | 3 - 4 | 1652 | 81 | |
| GenBank mRNA | AY039605.1 | - | 3 - 4 | 1466 | 81 | |
| BT003021.1 | - | 2 - 3 | 1188 | 81 | ||
More Information
Set Membership: |
|

AT3G50910
Genomic Map