Assay Details
| Gene Symbol: MUR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114976.4 | NP_190685.2 | 1 | 426 | 76 | |
| GenBank mRNA | BT025710.1 | - | 1 | 55 | 76 | |
| U81805.1 | - | 1 | 103 | 76 | ||
More Information
Set Membership: |
|

MUR1
Genomic Map