Assay Details
| Gene Symbol: ROP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114989.4 | NP_190698.1 | 5 - 6 | 836 | 68 | |
| GenBank mRNA | AF375412.1 | - | 5 - 6 | 800 | 68 | |
| AY066054.1 | - | 5 - 6 | 497 | 68 | ||
| Gene Symbol: AT3G51290 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203123.1 | NP_001190052.1 | 8 - 9 | 2768 | 68 | |
More Information
Set Membership: |
|

ROP1
Genomic Map