Assay Details
| Gene Symbol: AT4G09580 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117026.5 | NP_192696.3 | 2 - 3 | 701 | 74 | |
| GenBank mRNA | AY096649.1 | - | 2 - 3 | 632 | 74 | |
| AY114042.1 | - | 2 - 3 | 560 | 74 | ||
More Information
Set Membership: |
|

AT4G09580
Genomic Map