Assay Details
| Gene Symbol: AT4G09810 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340637.1 | NP_001328904.1 | 4 - 5 | 933 | 87 | |
| NM_117049.5 | NP_192719.1 | 5 - 6 | 882 | 87 | ||
| GenBank mRNA | BT010950.1 | - | 4 - 5 | 731 | 87 | |
| KJ667161.1 | - | 4 - 5 | 683 | 87 | ||
| KP872772.1 | - | 4 - 5 | 683 | 87 | ||
More Information
Set Membership: |
|

AT4G09810
Genomic Map