Assay Details
| Gene Symbol: STK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084895.1 | NP_001078364.1 | 4 - 5 | 593 | 112 | |
| NM_001340645.1 | NP_001319889.1 | 5 - 6 | 774 | 112 | ||
| NM_001340646.1 | NP_001329611.1 | 4 - 5 | 1059 | 112 | ||
| NM_001340647.1 | NP_001329612.1 | 4 - 5 | 525 | 112 | ||
| NM_117064.7 | NP_192734.1 | 4 - 5 | 564 | 112 | ||
| NM_179020.5 | NP_849351.1 | 4 - 5 | 411 | 112 | ||
| GenBank mRNA | AY065289.1 | - | 4 - 5 | 368 | 112 | |
| AY087201.1 | - | 4 - 5 | 471 | 112 | ||
| AY133738.1 | - | 3 - 4 | 328 | 112 | ||
| U20182.1 | - | 3 - 4 | 347 | 112 | ||
More Information
Set Membership: |
|

STK
Genomic Map