Assay Details
| Gene Symbol: STK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084895.1 | NP_001078364.1 | 5 - 6 | 691 | 82 | |
| NM_001340645.1 | NP_001319889.1 | 6 - 7 | 872 | 82 | ||
| NM_001340646.1 | NP_001329611.1 | 5 - 6 | 1157 | 82 | ||
| NM_001340647.1 | NP_001329612.1 | 5 - 6 | 623 | 82 | ||
| NM_117064.7 | NP_192734.1 | 5 - 6 | 662 | 82 | ||
More Information
Set Membership: |
|

STK
Genomic Map