Assay Details
| Gene Symbol: CID12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036537.1 | NP_001031614.1 | 6 - 7 | 931 | 93 | |
| NM_117129.3 | NP_192799.1 | 6 - 7 | 1126 | 93 | ||
| GenBank mRNA | AY039599.1 | - | 6 - 7 | 870 | 93 | |
| AY078011.1 | - | 6 - 7 | 772 | 93 | ||
| AY086286.1 | - | 6 - 7 | 848 | 93 | ||
More Information
Set Membership: |
|

CID12
Genomic Map