Assay Details
| Gene Symbol: MTSSB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117176.3 | NP_192844.2 | 6 - 7 | 820 | 111 | |
| GenBank mRNA | AJ582222.1 | - | 5 - 6 | 592 | 111 | |
| BT004166.1 | - | 6 - 7 | 757 | 111 | ||
| BT005015.1 | - | 5 - 6 | 592 | 111 | ||
More Information
Set Membership: |
|

MTSSB
Genomic Map